Assay Details
Gene Symbol: ACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100030.4 | NP_171655.1 | 3 - 4 | 619 | 64 | |
NM_179241.1 | NP_849572.1 | 2 - 3 | 358 | 64 | ||
GenBank mRNA | AF334719.1 | - | 3 - 4 | 463 | 64 | |
AY052207.1 | - | 2 - 3 | 358 | 64 | ||
AY143877.1 | - | 1 - 2 | 145 | 64 | ||
M95595.1 | - | 3 - 4 | 463 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
