Assay Details
Gene Symbol: LYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102036.4 | NP_564153.1 | 1 - 2 | 966 | 91 | |
NM_179366.1 | NP_849697.1 | 1 - 2 | 875 | 91 | ||
GenBank mRNA | AY057542.1 | - | 1 - 2 | 875 | 91 | |
BT015782.1 | - | 1 - 2 | 667 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
