Assay Details
Gene Symbol: AT3G13610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112207.4 | NP_187970.1 | 1 - 2 | 702 | 86 | |
GenBank mRNA | AK221576.1 | - | 1 - 2 | 587 | 86 | |
BT011745.1 | - | 1 - 2 | 513 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
