Assay Details
Gene Symbol: MYB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112243.3 | NP_566467.2 | 1 - 2 | 164 | 140 | |
GenBank mRNA | AF175997.1 | - | 1 - 2 | 164 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
