Assay Details
Gene Symbol: MYB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084678.2 | NP_001078147.1 | 2 - 3 | 312 | 116 | |
NM_112243.3 | NP_566467.2 | 2 - 3 | 284 | 116 | ||
GenBank mRNA | AF175997.1 | - | 2 - 3 | 284 | 116 | |
AY519588.1 | - | 2 - 3 | 257 | 116 | ||
DQ446661.1 | - | 2 - 3 | 257 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
