Assay Details
Gene Symbol: AT3G14910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112353.4 | NP_188109.1 | 2 - 3 | 504 | 85 | |
GenBank mRNA | BT022104.1 | - | 2 - 3 | 489 | 85 | |
BT029181.1 | - | 2 - 3 | 483 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
