Assay Details
Gene Symbol: AT3G14910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112353.4 | NP_188109.1 | 7 - 8 | 1201 | 56 | |
GenBank mRNA | BT022104.1 | - | 7 - 8 | 1186 | 56 | |
BT029181.1 | - | 7 - 8 | 1180 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
