Assay Details
Gene Symbol: CCD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342177.1 | NP_001329787.1 | 1 - 2 | 434 | 73 | |
NM_119434.4 | NP_195007.2 | 1 - 2 | 434 | 73 | ||
GenBank mRNA | AY074264.1 | - | 1 - 2 | 430 | 73 | |
AY133732.1 | - | 1 - 2 | 314 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
