Assay Details
Gene Symbol: EDA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119583.4 | NP_195146.1 | 3 - 4 | 1244 | 88 | |
GenBank mRNA | AY063810.1 | - | 3 - 4 | 1200 | 88 | |
AY150462.1 | - | 3 - 4 | 1139 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
