Assay Details
Gene Symbol: AT4G34290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119594.3 | NP_195155.1 | 1 - 2 | 407 | 75 | |
GenBank mRNA | BT001198.1 | - | 1 - 2 | 315 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
