Assay Details
Gene Symbol: MSI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119671.3 | NP_195231.1 | 2 - 3 | 828 | 110 | |
GenBank mRNA | BT020491.1 | - | 2 - 3 | 681 | 110 | |
BT025992.1 | - | 2 - 3 | 651 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
