Assay Details
Gene Symbol: MSI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119671.3 | NP_195231.1 | 3 - 4 | 961 | 84 | |
GenBank mRNA | BT020491.1 | - | 3 - 4 | 814 | 84 | |
BT025992.1 | - | 3 - 4 | 784 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
