Assay Details
Gene Symbol: AT1G69060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179533.3 | NP_849864.1 | 2 - 3 | 1867 | 89 | |
GenBank mRNA | AY091020.1 | - | 2 - 3 | 1604 | 89 | |
AY133814.1 | - | 1 - 2 | 1479 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
