Assay Details
Gene Symbol: AT1G69060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179533.3 | NP_849864.1 | 4 - 5 | 2134 | 93 | |
GenBank mRNA | AY091020.1 | - | 4 - 5 | 1871 | 93 | |
AY133814.1 | - | 3 - 4 | 1746 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
