Assay Details
Gene Symbol: AT2G17760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179636.3 | NP_849967.1 | 1 - 2 | 555 | 70 | |
GenBank mRNA | AY069886.1 | - | 1 - 2 | 441 | 70 | |
AY142012.1 | - | 1 - 2 | 301 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
