Assay Details
Gene Symbol: AT2G17760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179636.3 | NP_849967.1 | 9 - 10 | 1600 | 79 | |
GenBank mRNA | AY069886.1 | - | 9 - 10 | 1486 | 79 | |
AY142012.1 | - | 9 - 10 | 1346 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
