Assay Details
Gene Symbol: AT2G24395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179721.4 | NP_850052.1 | 2 - 3 | 373 | 87 | |
GenBank mRNA | AK221846.1 | - | 2 - 3 | 342 | 87 | |
BT011725.1 | - | 2 - 3 | 315 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
