Assay Details
Gene Symbol: HDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336129.1 | NP_001318300.1 | 2 - 3 | 172 | 123 | |
NM_001336130.1 | NP_001324791.1 | 2 - 3 | 145 | 123 | ||
NM_128344.4 | NP_565661.2 | 2 - 3 | 360 | 123 | ||
NM_179778.1 | NP_850109.1 | 1 - 2 | 82 | 123 | ||
GenBank mRNA | AF255713.1 | - | 1 - 2 | 81 | 123 | |
AK229619.1 | - | 2 - 3 | 145 | 123 | ||
BT006162.1 | - | 2 - 3 | 145 | 123 | ||
BT008535.1 | - | 1 - 2 | 81 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_179778.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
