Assay Details
Gene Symbol: PARG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336350.1 | NP_001324969.1 | 2 - 3 | 533 | 72 | |
NM_179844.3 | NP_850175.1 | 2 - 3 | 533 | 72 | ||
NM_201849.3 | NP_973578.1 | 2 - 3 | 533 | 72 | ||
GenBank mRNA | AY072330.1 | - | 2 - 3 | 498 | 72 | |
BT010379.1 | - | 2 - 3 | 434 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
