Assay Details
Gene Symbol: AT3G15357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112403.2 | NP_566510.1 | 1 | 366 | 61 | |
GenBank mRNA | AY085841.1 | - | 1 | 330 | 61 | |
BT024820.1 | - | 1 | 284 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
