Assay Details
Gene Symbol: AT3G16370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112508.5 | NP_188258.1 | 3 - 4 | 824 | 86 | |
GenBank mRNA | AY075698.1 | - | 3 - 4 | 630 | 86 | |
BT002609.1 | - | 3 - 4 | 606 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
