Assay Details
Gene Symbol: TPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112557.4 | NP_188306.2 | 12 - 13 | 1631 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
