Assay Details
Gene Symbol: ENA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084702.2 | NP_001078171.1 | 2 - 3 | 458 | 73 | |
NM_001338302.1 | NP_001327029.1 | 2 - 3 | 458 | 73 | ||
GenBank mRNA | AK176588.1 | - | 2 - 3 | 253 | 73 | |
AK176702.1 | - | 2 - 3 | 263 | 73 | ||
AK176807.1 | - | 2 - 3 | 263 | 73 | ||
BT015109.1 | - | 2 - 3 | 226 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
