Assay Details
Gene Symbol: AT3G17670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338303.1 | NP_001319578.1 | 1 - 2 | 444 | 64 | |
NM_112648.3 | NP_188394.2 | 1 - 2 | 464 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
