Assay Details
Gene Symbol: AT3G17670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338303.1 | NP_001319578.1 | 2 - 3 | 490 | 72 | |
NM_112648.3 | NP_188394.2 | 2 - 3 | 510 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
