Assay Details
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035654.2 | NP_001030731.1 | 12 - 13 | 1224 | 69 | |
NM_001338474.1 | NP_001327849.1 | 13 - 14 | 1302 | 69 | ||
NM_112920.4 | NP_566657.1 | 13 - 14 | 1369 | 69 | ||
GenBank mRNA | AK317308.1 | - | 12 - 13 | 1290 | 69 | |
AY034911.1 | - | 13 - 14 | 1226 | 69 | ||
BT004414.1 | - | 12 - 13 | 1096 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
