Assay Details
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035654.2 | NP_001030731.1 | 13 - 14 | 1309 | 101 | |
NM_001338474.1 | NP_001327849.1 | 14 - 15 | 1387 | 101 | ||
NM_112920.4 | NP_566657.1 | 14 - 15 | 1454 | 101 | ||
GenBank mRNA | AK317308.1 | - | 13 - 14 | 1375 | 101 | |
AY034911.1 | - | 14 - 15 | 1311 | 101 | ||
BT004414.1 | - | 13 - 14 | 1181 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
