Assay Details
Gene Symbol: LTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342367.1 | NP_001329911.1 | 5 - 6 | 960 | 127 | |
NM_119742.4 | NP_567988.1 | 5 - 6 | 960 | 127 | ||
GenBank mRNA | AY099728.1 | - | 5 - 6 | 946 | 127 | |
AY128903.1 | - | 5 - 6 | 905 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
