Assay Details
Gene Symbol: LTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119742.4 | NP_567988.1 | 6 - 7 | 1099 | 84 | |
GenBank mRNA | AY099728.1 | - | 6 - 7 | 1085 | 84 | |
AY128903.1 | - | 6 - 7 | 1044 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
