Assay Details
Gene Symbol: AT4G36430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119806.3 | NP_195361.1 | 2 - 3 | 533 | 81 | |
GenBank mRNA | AF452384.1 | - | 2 - 3 | 454 | 81 | |
AJ286345.1 | - | 2 - 3 | 418 | 81 | ||
AY074296.1 | - | 2 - 3 | 466 | 81 | ||
AY117238.1 | - | 2 - 3 | 417 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
