Assay Details
Gene Symbol: MES9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119878.5 | NP_195432.1 | 2 - 3 | 706 | 108 | |
GenBank mRNA | AB493724.1 | - | 2 - 3 | 484 | 108 | |
AK117107.1 | - | 2 - 3 | 534 | 108 | ||
BT006227.1 | - | 2 - 3 | 484 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
