Assay Details
Gene Symbol: AT4G38370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119999.3 | NP_568040.1 | 3 - 4 | 379 | 73 | |
GenBank mRNA | AK226503.1 | - | 3 - 4 | 342 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
