Assay Details
Gene Symbol: AT4G38370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119999.3 | NP_568040.1 | 5 - 6 | 570 | 78 | |
GenBank mRNA | AK226503.1 | - | 5 - 6 | 533 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
