Assay Details
Gene Symbol: HTR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035850.3 | NP_001030927.1 | 2 - 3 | 378 | 108 | |
NM_001331269.1 | NP_001322867.1 | 3 - 4 | 291 | 108 | ||
NM_001331270.1 | NP_001322868.1 | 3 - 4 | 261 | 108 | ||
NM_100019.2 | NP_563627.1 | 3 - 4 | 291 | 108 | ||
GenBank mRNA | AB081500.1 | - | 2 - 3 | 103 | 108 | |
AF465800.1 | - | 2 - 3 | 94 | 108 | ||
AK175840.1 | - | 2 - 3 | 150 | 108 | ||
BT024771.1 | - | 2 - 3 | 94 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
