Assay Details
Gene Symbol: ACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100030.4 | NP_171655.1 | 1 - 2 | 323 | 98 | |
GenBank mRNA | AF334719.1 | - | 1 - 2 | 167 | 98 | |
M95595.1 | - | 1 - 2 | 167 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
