Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 2 - 3 | 432 | 140 | |
GenBank mRNA | AF370292.1 | - | 2 - 3 | 353 | 140 | |
AK176092.1 | - | 2 - 3 | 382 | 140 | ||
AY063040.1 | - | 2 - 3 | 341 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
