Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 3 - 4 | 528 | 113 | |
GenBank mRNA | AF370292.1 | - | 3 - 4 | 449 | 113 | |
AK176092.1 | - | 3 - 4 | 478 | 113 | ||
AY063040.1 | - | 3 - 4 | 437 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
