Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 4 - 5 | 582 | 108 | |
GenBank mRNA | AF370292.1 | - | 4 - 5 | 503 | 108 | |
AK176092.1 | - | 4 - 5 | 532 | 108 | ||
AY063040.1 | - | 4 - 5 | 491 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
