Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 5 - 6 | 622 | 83 | |
GenBank mRNA | AF370292.1 | - | 5 - 6 | 543 | 83 | |
AK176092.1 | - | 5 - 6 | 572 | 83 | ||
AY063040.1 | - | 5 - 6 | 531 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
