Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 6 - 7 | 678 | 99 | |
GenBank mRNA | AF370292.1 | - | 6 - 7 | 599 | 99 | |
AK176092.1 | - | 6 - 7 | 628 | 99 | ||
AY063040.1 | - | 6 - 7 | 587 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
