Assay Details
Gene Symbol: AT1G02475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100129.4 | NP_563656.1 | 7 - 8 | 745 | 88 | |
GenBank mRNA | AF370292.1 | - | 7 - 8 | 666 | 88 | |
AK176092.1 | - | 7 - 8 | 695 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
