Assay Details
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100148.3 | NP_171768.2 | 5 - 6 | 410 | 78 | |
GenBank mRNA | AB493426.1 | - | 4 - 5 | 275 | 78 | |
AK175906.1 | - | 5 - 6 | 410 | 78 | ||
AK175966.1 | - | 5 - 6 | 410 | 78 | ||
AK176854.1 | - | 5 - 6 | 378 | 78 | ||
AK221393.1 | - | 5 - 6 | 389 | 78 | ||
AY463615.1 | - | 4 - 5 | 275 | 78 | ||
BT010482.1 | - | 4 - 5 | 275 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
