Assay Details
Gene Symbol: NET1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331385.1 | NP_001322259.1 | 3 - 4 | 606 | 102 | |
NM_001331386.1 | NP_001322258.1 | 2 - 3 | 569 | 102 | ||
NM_100190.4 | NP_171807.2 | 3 - 4 | 695 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
