Assay Details
Gene Symbol: TPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100294.3 | NP_563702.1 | 1 - 2 | 292 | 120 | |
GenBank mRNA | AY054170.1 | - | 1 - 2 | 138 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
