Assay Details
Gene Symbol: emb2411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198014.1 | NP_001184943.1 | 26 - 27 | 3240 | 109 | |
NM_001331794.1 | NP_001318959.1 | 27 - 28 | 3345 | 109 | ||
NM_001331795.1 | NP_001322072.1 | 28 - 29 | 3446 | 109 | ||
NM_001331796.1 | NP_001322073.1 | 28 - 29 | 3440 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
