Assay Details
Gene Symbol: AT1G09390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100809.5 | NP_172410.1 | 2 - 3 | 658 | 87 | |
GenBank mRNA | BT002885.1 | - | 2 - 3 | 476 | 87 | |
BT004397.1 | - | 2 - 3 | 440 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
