Assay Details
Gene Symbol: AT1G09390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100809.5 | NP_172410.1 | 4 - 5 | 1104 | 114 | |
GenBank mRNA | BT002885.1 | - | 4 - 5 | 922 | 114 | |
BT004397.1 | - | 4 - 5 | 886 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
