Assay Details
Gene Symbol: ATP-PRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100852.2 | NP_563853.1 | 10 - 11 | 1204 | 96 | |
GenBank mRNA | AK118503.1 | - | 10 - 11 | 1190 | 96 | |
BT002054.1 | - | 10 - 11 | 1190 | 96 | ||
BT008501.1 | - | 10 - 11 | 1150 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
