Assay Details
Gene Symbol: NDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125058.3 | NP_200486.1 | 1 - 2 | 420 | 70 | |
GenBank mRNA | AY050428.1 | - | 1 - 2 | 172 | 70 | |
AY087544.1 | - | 1 - 2 | 172 | 70 | ||
AY090332.1 | - | 1 - 2 | 63 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
