Assay Details
Gene Symbol: NDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345216.1 | NP_001330516.1 | 2 - 3 | 400 | 118 | |
NM_125058.3 | NP_200486.1 | 2 - 3 | 516 | 118 | ||
GenBank mRNA | AY050428.1 | - | 2 - 3 | 268 | 118 | |
AY087544.1 | - | 2 - 3 | 118 | |||
AY090332.1 | - | 2 - 3 | 159 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
